missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dimethyl pyrimidine-2,5-dicarboxylate, 97%

Brand:  Alfa Aesar™ H64903.06

Code : NEW

Additional Details : CAS Number : 115600-17-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15482309

  • £201.40 / 5g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

Dimethyl pyrimidine-2,5-dicarboxylate
196.16
23483097
115600-17-4
MFCD26127433
Pyrimidine-2,5-dicarboxylic acid dimethyl ester

Specifications

Dimethyl pyrimidine-2,5-dicarboxylate
5g
Powder
196.16
23483097
White
115600-17-4
97%
MFCD26127433
Pyrimidine-2,5-dicarboxylic acid dimethyl ester
Videos
SDS