missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Phthalate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.1900g/mL |
Boiling Point | 282°C |
Flash Point | 150°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Metal drum |
Refractive Index | 1.5140 to 1.516 |
Quantity | 25 L |
Beilstein | 09, 797 |
Merck Index | 15, 3279 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'