missing translation for 'onlineSavingsMsg'
Learn More

α,α-Dimethyl-β-methylsuccinimide, 99+%, ACROS Organics™

Brand:  Acros Organics 172512500

Code : 29

Additional Details : CAS Number : 61748-86-5 Weight : 0.05025kg

Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10205911

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

α, α-Dimethyl-β-methylsuccinimide
≥99%
C7H11NO2
MFCD00005497
61748-86-5
99+%
141.17

Specifications

α, α-Dimethyl-β-methylsuccinimide
117.0° to 122.0°C
61748-86-5
Glass bottle
≥99%
C7H11NO2
MFCD00005497
White
250mg
Authentic
Crystalline Powder
99+%
141.17
21, 394
Videos
SDS