missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 1.1400g/mL |
Boiling Point | 181.0°C |
Flash Point | 68°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3O)2P(O)CH3 |
Refractive Index | 1.413 |
Quantity | 5 g |
Show More |
Safety and Handling
Product Identifier
- Dimethyl methylphosphonate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- H340-May cause genetic defects.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'