missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl methoxymethylenemalonate, 98+%, Thermo Scientific Chemicals

Catalog Number p-118307
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
22398-14-7
C7H10O5
174.16
MFCD00038225
RHFZTBSULNJWEI-UHFFFAOYSA-N
dimethyl methoxymethylenemalonate, dimethyl 2-methoxymethylene malonate, 1,3-dimethyl 2-methoxymethylidene propanedioate, methoxymethylenemalonic acid dimethyl ester, dimethyl methoxymethylene malonate, acmc-1cq9e, dimethylmethoxymethylenemalonate, propanedioic acid, methoxymethylene-, dimethyl ester, dimethyl methoxymethylidenemalonate
90770
dimethyl 2-(methoxymethylidene)propanedioate
COC=C(C(=O)OC)C(=O)OC
This item is not returnable. View return policy
22398-14-7
C7H10O5
174.16
MFCD00038225
RHFZTBSULNJWEI-UHFFFAOYSA-N
dimethyl methoxymethylenemalonate, dimethyl 2-methoxymethylene malonate, 1,3-dimethyl 2-methoxymethylidene propanedioate, methoxymethylenemalonic acid dimethyl ester, dimethyl methoxymethylene malonate, acmc-1cq9e, dimethylmethoxymethylenemalonate, propanedioic acid, methoxymethylene-, dimethyl ester, dimethyl methoxymethylidenemalonate
90770
dimethyl 2-(methoxymethylidene)propanedioate
COC=C(C(=O)OC)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 40°C to 44°C
Color White
Boiling Point 98°C (0.5mmHg)
Flash Point 158°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH=C(COOCH3)2
Quantity 25 g
Solubility Information Solubility in water: may hydrolyse. Other solubilities: soluble in methanol 1260g/L (20 c)
Formula Weight 174.16
Percent Purity 98+%
Physical Form Crystalline Solid
Chemical Name or Material Dimethyl methoxymethylenemalonate, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl methoxymethylenemalonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'