Learn More
Dimethyl malonate, 98+%, Thermo Scientific Chemicals
Description
Dimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.
Solubility
Miscible with alcohol, ether and oils. Slightly miscible with water.
Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications
Melting Point | -62°C |
Density | 1.156 |
Boiling Point | 180°C to 181°C |
Flash Point | 90°C (194°F) |
Refractive Index | 1.413 |
Quantity | 100 g |
Beilstein | 774261 |
Merck Index | 14,6096 |
Solubility Information | Miscible with alcohol,ether and oils. Slightly miscible with water. |
Formula Weight | 132.12 |
Show More |
RUO – Research Use Only