missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl isobutylmalonate, 98%, ACROS Organics™

Brand:  Acros Organics 350470250

Code : 29

Additional Details : CAS Number : 2544-90-8 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10673563

  • £94.60 / 25g


Estimated Shipment
18-05-2018
Add to basket

Specifications

Chemical Identifiers

Dimethyl isobutylmalonate
39520-24-6
98%
188.23
98%
98%
C9H16O4
MFCD00015640

Specifications

Dimethyl isobutylmalonate
Clear colorless to light yellow
98%
Authentic
1.4230 to 1.4250
98%
C9H16O4
MFCD00015640
211.0°C (763.0 mmHg)
25g
39520-24-6
Glass bottle
Liquid
98%
188.23
Videos
SDS