missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl isobutylmalonate, 98%, Thermo Scientific™

Catalog Number 10232802
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
39520-24-6
C9H16O4
188.22
MFCD00015640
GAZFDPSEEIVCEX-UHFFFAOYSA-N
dimethyl isobutylmalonate, dimethyl 2-isobutylmalonate, dimethylisobutylmalonate, propanedioic acid, 2-2-methylpropyl-, 1,3-dimethyl ester, propanedioic acid, 2-methylpropyl-, dimethyl ester, 1,3-dimethyl 2-2-methylpropyl propanedioate, malonic acid, isobutyl-, dimethyl ester, acmc-1ailj, dimethyl2-isobutylmalonate
170218
dimethyl 2-(2-methylpropyl)propanedioate
COC(=O)C(CC(C)C)C(=O)OC
This item is not returnable. View return policy
39520-24-6
C9H16O4
188.22
MFCD00015640
GAZFDPSEEIVCEX-UHFFFAOYSA-N
dimethyl isobutylmalonate, dimethyl 2-isobutylmalonate, dimethylisobutylmalonate, propanedioic acid, 2-2-methylpropyl-, 1,3-dimethyl ester, propanedioic acid, 2-methylpropyl-, dimethyl ester, 1,3-dimethyl 2-2-methylpropyl propanedioate, malonic acid, isobutyl-, dimethyl ester, acmc-1ailj, dimethyl2-isobutylmalonate
170218
dimethyl 2-(2-methylpropyl)propanedioate
COC(=O)C(CC(C)C)C(=O)OC

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point 211.0°C (763.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4230 to 1.4250
Quantity 5 g
Formula Weight 188.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dimethyl isobutylmalonate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'