missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl diselenide, 99%, Thermo Scientific Chemicals
47.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
Dimethyl diselenide is a powerful reagent used for identification of distonic radical cations
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.987 g/mL |
Boiling Point | 155°C to 157°C |
Flash Point | 60°C (140°F) |
Odor | Unpleasant |
Refractive Index | 1.6388 |
Quantity | 1 g |
UN Number | UN2929 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 187.99 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'