missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cyanodithioiminocarbonate, 95%, Thermo Scientific Chemicals

Catalog Number p-7081183
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
10191-60-3
C4H6N2S2
146.226
MFCD00009825
IULFXBLVJIPESI-UHFFFAOYSA-N
dimethyl cyanocarbonimidodithioate, dimethyl cyanodithioiminocarbonate, dimethyl n-cyanodithioiminocarbonate, carbonimidodithioic acid, cyano-, dimethyl ester, s,s-dimethyl cyanoimidodithiocarbonate, cyanoimino bis methylthio methane, dimethyl n-cyanoimidodithiocarbonate, dimethyl cyanimidodithiocarbonate, dimethyl cyanoiminodithiocarbonate, dimethyl n-cyanodithiocarbonimidate
66289
bis(methylsulfanyl)methylidenecyanamide
CSC(=NC#N)SC
This item is not returnable. View return policy
10191-60-3
C4H6N2S2
146.226
MFCD00009825
IULFXBLVJIPESI-UHFFFAOYSA-N
dimethyl cyanocarbonimidodithioate, dimethyl cyanodithioiminocarbonate, dimethyl n-cyanodithioiminocarbonate, carbonimidodithioic acid, cyano-, dimethyl ester, s,s-dimethyl cyanoimidodithiocarbonate, cyanoimino bis methylthio methane, dimethyl n-cyanoimidodithiocarbonate, dimethyl cyanimidodithiocarbonate, dimethyl cyanoiminodithiocarbonate, dimethyl n-cyanodithiocarbonimidate
66289
bis(methylsulfanyl)methylidenecyanamide
CSC(=NC#N)SC

Dimethyl cyanodithioiminocarbonate has been used in the synthesis of 4-methylthiopyrazolo[1,5-a]-1,3,5-triazines, methylsulfanylpyrimidines, cyanoguanidines and N-aryl-6-methylsulfanyl-4-oxopyrimidine-5-carbonitriles. It has been also used in the synthesis of methylsulfanyl derivatives of azoloazines and azoloazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl cyanodithioiminocarbonate has been used in the synthesis of 4-methylthiopyrazolo[1,5-a]-1,3,5-triazines, methylsulfanylpyrimidines, cyanoguanidines and N-aryl-6-methylsulfanyl-4-oxopyrimidine-5-carbonitriles. It has been also used in the synthesis of methylsulfanyl derivatives of azoloazines and azoloazoles.

Solubility
Slightly soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 50°C to 56°C
Flash Point >110°C (230°F)
Odor Stench
Linear Formula NCN=C(SCH3)2
Quantity 25 g
UN Number UN2923
Beilstein 635998
Solubility Information Slightly soluble in methanol.
Formula Weight 146.23
Percent Purity 95%
Chemical Name or Material Dimethyl cyanodithioiminocarbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'