missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl carbonate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4089508
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
616-38-6
C3H6O3
90.08
MFCD00008420
IEJIGPNLZYLLBP-UHFFFAOYSA-N
methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
12021
CHEBI:36596
dimethyl carbonate
COC(=O)OC
616-38-6
C3H6O3
90.08
MFCD00008420
IEJIGPNLZYLLBP-UHFFFAOYSA-N
methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
12021
CHEBI:36596
dimethyl carbonate
COC(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2.0°C to 4.0°C
Color Brown to White
Density 1.0690g/mL
Boiling Point 90.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3O)2CO
Refractive Index 1.3672 to 1.3692
Quantity 100 mL
Merck Index 15, 3263
Specific Gravity 1.069
Solubility Information Solubility in water: 139g/L. Other solubilities: soluble in all proportions with organic alcohols,soluble in all proportions with esters and ketones,miscible with acids and alkalies
Viscosity 0.625 mPa.s (20°C)
Formula Weight 90.08
Percent Purity 99+%
Grade Extra Dry
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl carbonate, Extra Dry
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl carbonate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'