missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl aminoterephthalate, 99%, Thermo Scientific Chemicals

Catalog Number 10624892
Click to view available options
Quantity:
25 g
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
5372-81-6
C10H11NO4
209.2
MFCD00008427
DSSKDXUDARIMTR-UHFFFAOYSA-N
dimethyl 2-aminoterephthalate, dimethyl aminoterephthalate, dimethyl 3-aminoterephthalate, 1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester, aminoterephthalic acid dimethyl ester, 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate, unii-91sf4e6i9w, 2-aminoterephthalic acid, dimethyl ester, 3-amino-4-methoxycarbonylbenzoic acid, methyl ester, terephthalic acid, amino-, dimethyl ester
79336
dimethyl 2-aminobenzene-1,4-dicarboxylate
COC(=O)C1=CC(=C(C=C1)C(=O)OC)N
This item is not returnable. View return policy
5372-81-6
C10H11NO4
209.2
MFCD00008427
DSSKDXUDARIMTR-UHFFFAOYSA-N
dimethyl 2-aminoterephthalate, dimethyl aminoterephthalate, dimethyl 3-aminoterephthalate, 1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester, aminoterephthalic acid dimethyl ester, 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate, unii-91sf4e6i9w, 2-aminoterephthalic acid, dimethyl ester, 3-amino-4-methoxycarbonylbenzoic acid, methyl ester, terephthalic acid, amino-, dimethyl ester
79336
dimethyl 2-aminobenzene-1,4-dicarboxylate
COC(=O)C1=CC(=C(C=C1)C(=O)OC)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 132.0°C to 134.0°C
Color Beige or Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula H2NC6H3(CO2CH3)2
Quantity 250 g
Beilstein 14, 559
Formula Weight 209.2
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material Dimethyl aminoterephthalate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl aminoterephthalate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'