missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl allylmalonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Notes
Protect from moisture, heat. Incomaptible with oxidizing agents. Store in cool place.
Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Notes
Protect from moisture, heat. Incomaptible with oxidizing agents. Store in cool place.
Specifications
Melting Point | <-80°C |
Density | 1.074 |
Boiling Point | 207°C |
Flash Point | 95°C (203°F) |
Refractive Index | 1.435 |
Quantity | 10 g |
Beilstein | 1772822 |
Formula Weight | 172.18 |
Percent Purity | 97% |
Chemical Name or Material | Dimethyl allylmalonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'