missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog Number p-7043385
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4202-14-6
C5H11O4P
166.113
MFCD00008769
UOWIYNWMROWVDG-UHFFFAOYSA-N
dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone
77872
1-dimethoxyphosphorylpropan-2-one
CC(=O)CP(=O)(OC)OC
This item is not returnable. View return policy
4202-14-6
C5H11O4P
166.113
MFCD00008769
UOWIYNWMROWVDG-UHFFFAOYSA-N
dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone
77872
1-dimethoxyphosphorylpropan-2-one
CC(=O)CP(=O)(OC)OC

Dimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.202
Boiling Point 121°C to 123°C (11 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.439
Quantity 5 g
Beilstein 1766218
Solubility Information Difficult to mix.
Formula Weight 166.12
Percent Purity 97%
Chemical Name or Material Dimethyl acetylmethylphosphonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'