missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethane, 99.5+%, Thermo Scientific Chemicals

Catalog Number 10498250
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
109-87-5
C3H8O2
76.09
MFCD00008495
NKDDWNXOKDWJAK-UHFFFAOYSA-N
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
8020
CHEBI:48341
dimethoxymethane
COCOC
109-87-5
C3H8O2
76.09
MFCD00008495
NKDDWNXOKDWJAK-UHFFFAOYSA-N
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
8020
CHEBI:48341
dimethoxymethane
COCOC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -105.0°C
Color Colorless
Density 0.8600g/mL
Boiling Point 41.0°C to 42.0°C
Flash Point −18°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2(OCH3)2
Refractive Index 1.3525 to 1.3545
Quantity 1 L
Fieser 01,671; 02,271; 06,374; 07,115
Merck Index 15, 6086
Specific Gravity 0.86
Solubility Information Solubility in water: 32.3g/100 mL (16°C). Other solubilities: miscible with alcohol,ether,oils,hydrocarbons,miscible with most common organic solvents,very soluble in acetone,benzene
Viscosity 3.25 mPa.s (20°C)
Formula Weight 76.09
Percent Purity 99.5+%
Physical Form Liquid
Chemical Name or Material Dimethoxymethane
Show More Show Less
compliance-icons
Product Identifier
  • Dimethoxymethane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'