missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxydimethylsilane, 95+%, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4229179
18.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
1112-39-6
C4H12O2Si
120.22
MFCD00025691
JJQZDUKDJDQPMQ-UHFFFAOYSA-N
dimethyldimethoxysilane, silane, dimethoxydimethyl, kbm 22, unii-a3qdb1rs1c, dimethyl dimethoxysilane, dimethoxy dimethyl silane, a3qdb1rs1c, dimethoxy-dimethylsilane, dimethyl dimethoxy silane, dimethyl dimethoxy silicane
66187
dimethoxydimethylsilane
CO[Si](C)(C)OC
This item is not returnable. View return policy
1112-39-6
C4H12O2Si
120.22
MFCD00025691
JJQZDUKDJDQPMQ-UHFFFAOYSA-N
dimethyldimethoxysilane, silane, dimethoxydimethyl, kbm 22, unii-a3qdb1rs1c, dimethyl dimethoxysilane, dimethoxy dimethyl silane, a3qdb1rs1c, dimethoxy-dimethylsilane, dimethyl dimethoxy silane, dimethyl dimethoxy silicane
66187
dimethoxydimethylsilane
CO[Si](C)(C)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -80.0°C
Color Colorless to Yellow
Density 0.8650g/mL
Boiling Point 81.0°C to 82.0°C
Flash Point −8°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3)2Si(OCH3)2
Refractive Index 1.3690 to 1.3710
Quantity 100 mL
Specific Gravity 0.865
Solubility Information Solubility in water: reacts with water to form methanol
Formula Weight 120.23
Percent Purity 95+%
Physical Form Liquid
Chemical Name or Material Dimethoxydimethylsilane
Show More Show Less
compliance-icons
Product Identifier
  • Dimethoxydimethylsilane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'