missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine dichloroacetate, 95%, Thermo Scientific Chemicals

Catalog Number 15437868
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
660-27-5
C8H17Cl2NO2
230.13
MFCD00868288
ILKBHIBYKSHTKQ-UHFFFAOYSA-N
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
12617
2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine
OC(=O)C(Cl)Cl.CC(C)NC(C)C
This item is not returnable. View return policy
660-27-5
C8H17Cl2NO2
230.13
MFCD00868288
ILKBHIBYKSHTKQ-UHFFFAOYSA-N
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
12617
2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine
OC(=O)C(Cl)Cl.CC(C)NC(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Linear Formula [(CH3)2CH]2NH·Cl2CHCO2H
Quantity 1 g
Formula Weight 230.13
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Diisopropylamine dichloroacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'