missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl methylphosphonate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Diisopropyl methylphosphonate is used as a precursor to prepare beta-keto phosphonates. It acts as an intermediate for Horner-Wadsworth-Emmons olefination of carbonyl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diisopropyl methylphosphonate is used as a precursor to prepare beta-keto phosphonates. It acts as an intermediate for Horner-Wadsworth-Emmons olefination of carbonyl compounds.
Solubility
Miscible with water.
Notes
Store in cool place. Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, reducing agents, strong reducing agents and phosphorus halides.
Diisopropyl methylphosphonate is used as a precursor to prepare beta-keto phosphonates. It acts as an intermediate for Horner-Wadsworth-Emmons olefination of carbonyl compounds.
Solubility
Miscible with water.
Notes
Store in cool place. Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, reducing agents, strong reducing agents and phosphorus halides.
Specifications
Density | 0.976 |
Boiling Point | 50°C to 51°C (1 mmHg) |
Refractive Index | 1.41 |
Quantity | 5 g |
Beilstein | 1761635 |
Solubility Information | Miscible with water. |
Formula Weight | 180.18 |
Percent Purity | 95% |
Chemical Name or Material | Diisopropyl methylphosphonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'