missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl methylphosphonate, 90%, Thermo Scientific Chemicals

Catalog Number 13438658
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1445-75-6
C7H17O3P
180.18
MFCD00015022
WOAFDHWYKSOANX-UHFFFAOYSA-N
diisopropyl methylphosphonate, diisopropyl methanephosphonate, bis 1-methylethyl methylphosphonate, unii-56v3og5dc7, methylphosphonic acid diisopropyl ester, phosphonic acid, methyl-, bis 1-methylethyl ester, phosphonic acid, methyl-, diisopropyl ester, methylphosphonic acid bis 1-methylethyl ester, dsstox_cid_4051, dsstox_rid_77269
3073
CHEBI:77325
2-[methyl(propan-2-yloxy)phosphoryl]oxypropane
CC(C)OP(C)(=O)OC(C)C
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1445-75-6
C7H17O3P
180.18
MFCD00015022
WOAFDHWYKSOANX-UHFFFAOYSA-N
diisopropyl methylphosphonate, diisopropyl methanephosphonate, bis 1-methylethyl methylphosphonate, unii-56v3og5dc7, methylphosphonic acid diisopropyl ester, phosphonic acid, methyl-, bis 1-methylethyl ester, phosphonic acid, methyl-, diisopropyl ester, methylphosphonic acid bis 1-methylethyl ester, dsstox_cid_4051, dsstox_rid_77269
3073
CHEBI:77325
2-[methyl(propan-2-yloxy)phosphoryl]oxypropane
CC(C)OP(C)(=O)OC(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9850g/mL
Boiling Point 50.0°C to 51.0°C (1.0 mmHg)
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2C(H)O]2P(O)CH3
Quantity 25 g
Specific Gravity 0.985
Formula Weight 180.18
Percent Purity 90%
Chemical Name or Material Diisopropyl methylphosphonate
compliance-icons
Product Identifier
  • Diisopropyl methylphosphonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'