missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
21.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen), Radical initiators
Specifications
Melting Point | 3.0°C to 5.0°C |
Color | Orange-Yellow to Red |
Density | 1.0200g/mL |
Boiling Point | 75.0°C (0.2 mmHg) |
Flash Point | 106°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 92% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3)2CHO2CN=NCO2CH(CH3)2 |
Refractive Index | 1.4180 to 1.4220 |
Show More |
Safety and Handling
Product Identifier
- Diisopropyl azodicarboxylate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'