missing translation for 'onlineSavingsMsg'
Learn More

(+)-Diisopinocampheylchloroborane, 1.8M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 10553044
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
112246-73-8, 80-56-8, 71-43-2, 64742-89-8
C20H34BCl
320.75
MFCD00074808
PSEHHVRCDVOTID-NAVXHOJHSA-N
+-b-chlorodiisopinocamphenylborane
133124869
chloro-bis[(3R,4S,5R)-4,6,6-trimethyl-3-bicyclo[3.1.1]heptanyl]borane
C[C@H]1[C@@H]2C[C@H](C[C@@H]1B(Cl)[C@H]1C[C@H]3C[C@@H]([C@@H]1C)C3(C)C)C2(C)C
This item is not returnable. View return policy
112246-73-8, 80-56-8, 71-43-2, 64742-89-8
C20H34BCl
320.75
MFCD00074808
PSEHHVRCDVOTID-NAVXHOJHSA-N
+-b-chlorodiisopinocamphenylborane
133124869
chloro-bis[(3R,4S,5R)-4,6,6-trimethyl-3-bicyclo[3.1.1]heptanyl]borane
C[C@H]1[C@@H]2C[C@H](C[C@@H]1B(Cl)[C@H]1C[C@H]3C[C@@H]([C@@H]1C)C3(C)C)C2(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Packaging AcroSeal™ Glass Bottle
Quantity 800 mL
Solubility Information Solubility in water: reacts
Formula Weight 320.77
Chemical Name or Material (+)-Diisopinocampheylchloroborane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'