missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diisobutylaluminum hydride, 1M solution in hexane

Reducing agent

Brand:  Alfa Aesar™ 042589.AD

Code : NEW

Additional Details : CAS Number : 1191-15-7 Weight : 0.00050kg Transport : UN number : 3399 Chem class : 4.3 Pack group : I


Quantity 50mL

 View more versions of this product

Product Code. 15406495

  • £17.90 / 50mL


EU Stock 5
Add to basket


Specifications

Chemical Identifiers

Diisobutylaluminum hydride
[(CH3)2CHCH2]2AlH
MFCD00008928
1191-15-7
142.22
UN3399

Specifications

Diisobutylaluminum hydride
1191-15-7
−23°C (−10°F)
Colorless
Liquid
142.22
UN3399
1M solution in hexane
50mL
0.701g/mL
Air & Moisture Sensitive
[(CH3)2CHCH2]2AlH
MFCD00008928
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diisobutylaluminum hydride, 1M solution in hexane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 A
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains¬†: n-Hexane, Diisobutylaluminum hydride

SDS