missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl methylphosphonate, 97%, Thermo Scientific™

Catalog Number 11455470
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
7242-56-0
C142H142Fe4N11O8P9S4
2761.15
MFCD00015638
RQHGPKSXRBXJJL-UHFFFAOYNA-W
diisobutyl methylphosphonate, bis 2-methylpropyl methylphosphonate, diisobutyl methylphosphonate #, o,o-diisobutyl methylthiophosphonate, methylphosphonic acid diisobutyl ester, diisobutyl methylphosphonate 1g, methylphosphonic acid, di 2-methylpropyl ester, 1-isobutoxy methyl phosphoryl oxy-2-methyl-propane, 2-methyl-1-methyl 2-methylpropoxy phosphoryl oxypropane
566101
2-methyl-1-[methyl(2-methylpropoxy)phosphoryl]oxypropane
[Fe].[Fe].[Fe].[Fe].[C-]#N.[C-]#N.[C-]#N.CS(=O)(=O)C(C#N)C#N.CS(=O)(=O)C(C#N)C#N.CS(=O)(=O)C(C#N)C#N.CS(=O)(=O)C(C#N)C#N.CC(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)C[PH+](C1=CC=CC=C1)C1=CC=CC=C1.CC(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)C[PH+](C1=CC=CC=C1)C1=CC=CC=C1.CC(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)C[PH+](C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
7242-56-0
C142H142Fe4N11O8P9S4
2761.15
MFCD00015638
RQHGPKSXRBXJJL-UHFFFAOYNA-W
diisobutyl methylphosphonate, bis 2-methylpropyl methylphosphonate, diisobutyl methylphosphonate #, o,o-diisobutyl methylthiophosphonate, methylphosphonic acid diisobutyl ester, diisobutyl methylphosphonate 1g, methylphosphonic acid, di 2-methylpropyl ester, 1-isobutoxy methyl phosphoryl oxy-2-methyl-propane, 2-methyl-1-methyl 2-methylpropoxy phosphoryl oxypropane
566101
2-methyl-1-[methyl(2-methylpropoxy)phosphoryl]oxypropane
[Fe].[Fe].[Fe].[Fe].[C-]#N.[C-]#N.[C-]#N.CS(=O)(=O)C(C#N)C#N.CS(=O)(=O)C(C#N)C#N.CS(=O)(=O)C(C#N)C#N.CS(=O)(=O)C(C#N)C#N.CC(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)C[PH+](C1=CC=CC=C1)C1=CC=CC=C1.CC(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)C[PH+](C1=CC=CC=C1)C1=CC=CC=C1.CC(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)C[PH+](C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 10 g
Formula Weight 208.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diisobutyl methylphosphonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'