missing translation for 'onlineSavingsMsg'
Learn More

Dihydroxybis(ammonium lactato)titanium(IV), 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog Number p-7052148
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
65104-06-5
MFCD00072736
Titanium(IV) bis(ammonium lactato)dihydroxide
This item is not returnable. View return policy
65104-06-5
MFCD00072736
Titanium(IV) bis(ammonium lactato)dihydroxide

Water soluble titanium complex

Dihydroxybis(ammonium lactato)titanium(IV) is used in the production of water soluble titanium complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.21 g/mL
Flash Point 47°C (118°F)
Refractive Index 1.438
Quantity 250 g
UN Number UN1993
Solubility Information Miscible with water.
Formula Weight 294.1
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Dihydroxybis (ammonium lactato)titanium (IV)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'