Learn More
Dihydrokainic acid, Tocris Bioscience™
EAAT2 (GLT-1)-selective non-transportable inhibitor of L-glutamate and L-aspartate uptake
Brand:  RD Systems (Bio-Techne) 01111
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 1mg
 View more versions of this product
Product Code. 13487433
  • £86.80 / 1mg
Estimated Shipment
07-03-2018
Add to basket
Description
Description
EAAT2(GLT1)-selective non-transportable inhibitor of L-glutamate and L-aspartate uptake (Ki = 23 μM). 130-fold selective over EAAT1 and EAAT3 (Ki > 3 mM). Also available as part of the Excitatory Amino Acid Transporter Inhibitor Tocriset™ .
Specifications
Chemical Identifiers
Dihydrokainic acid
C10H17NO4
215.25
(2S, 3S, 4R)-2-Carboxy-4-isopropyl-3-pyrrolidineacetic acid
Specifications
Dihydrokainic acid
52497-36-6
C10H17NO4
(2S, 3S, 4R)-2-Carboxy-4-isopropyl-3-pyrrolidineacetic acid
1mg
>98%
215.25
Videos