missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dihydrogen hexachloroiridate(IV) hydrate, 99% (metals basis), Ir 38-42%

Brand:  Alfa Aesar™ 011031.14

Code : NEW

Additional Details : CAS Number : 110802-84-1 Weight : 0.02500kg Transport : UN number : 3260 Chem class : 1a Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11301658

  • £1146.00 / 25g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Dihydrogen hexachloroiridate(IV) hydrate
406.93 (Anhydrous)
UN3260
110802-84-1
MFCD00011328
Hydrogen hexachloroiridate(IV); Chloroiridic acid

Specifications

Dihydrogen hexachloroiridate(IV) hydrate
25g
Lumps
(metals basis)
406.93 (Anhydrous)
UN3260
Soluble in alcohol and water
Ir 38 to 42%
110802-84-1
99%
Black
MFCD00011328
Hydrogen hexachloroiridate(IV); Chloroiridic acid
Videos
SDS