Learn More
Thermo Scientific Alfa Aesar Dihydrogen hexabromoiridate(IV) hexahydrate, 99.9% (metals basis), Ir 24.1% min
Brand: Thermo Scientific Alfa Aesar 041009.03
Dihydrogen hexabromoiridate(IV) hexahydrate, 99.9% (metals basis), Ir 24.1% min
Code : NEW
Additional Details : Weight : 0.00100kg Transport : UN number : 3260 Chem class : 8 Pack group : III