missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dihydrogen hexabromoiridate(IV) hexahydrate, 99.9% (metals basis), Ir 24.1% min

Brand:  Alfa Aesar™ 041009.06

Code : NEW

Additional Details : Weight : 0.00500kg Transport : UN number : 3260 Chem class : 8 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11369407

  • £325.20 / 5g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

Dihydrogen hexabromoiridate(IV) hexahydrate
MFCD00798543
Bromoiridic Acid; Hydrogen hexabromoiridate(IV)
781.76 (673.67 Anhydrous)
UN3260

Specifications

Dihydrogen hexabromoiridate(IV) hexahydrate
5g
99.9%
781.76 (673.67 Anhydrous)
UN3260
Ir 24.1% min.
Powder
(metals basis)
MFCD00798543
Bromoiridic Acid; Hydrogen hexabromoiridate(IV)
Videos
SDS