missing translation for 'onlineSavingsMsg'
Learn More

Dihydro-β-erythroidine hydrobromide, Tocris Bioscience™

Inhibitor of α4β2, muscle type and Torepedo nAChRs

Brand:  Tocris (Bio-Techne) 234950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461084

  • £607.97 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Competitive nicotinic acetylcholine receptor antagonist with moderate selectivity for the neuronal α4 receptor subunit (IC50 values are 0.19 and 0.37 μM for α4β4 and α4β2 receptors respectively). Antagonizes behavioral effects of nicotine in vivo.
Specifications

Chemical Identifiers

29734-68-7
356.26
dihydro-beta-erythroidine hydrobromide, unii-ns1u03gc05, 2s,13bs-2-methoxy-2,3,5,6,8,9,10,13-octahydro-1h,12h-benzo i pyrano 3,4-g indolizin-12-one hydrobromide, dh?e, dihydro-?-erythroidine hydrobromide, 3, a-1,6-didehydro-14,17-dihydro-3-methoxy-16 15h-oxaerythrinan-15-one hydrobromide, 3beta-1,6-didehydro-14,17-dihydro-3-methoxy-16 15h-oxaerythrinan-15-one hydrobromide
COC1CC=C2CCN3C2(C1)C4=C(CC3)COC(=O)C4.Br
C16H22BrNO3
GFIGWAJEIMHJJB-LINSIKMZSA-N
11957537

Specifications

Dihydro-β-erythroidine hydrobromide
29734-68-7
dihydro-beta-erythroidine hydrobromide, unii-ns1u03gc05, 2s,13bs-2-methoxy-2,3,5,6,8,9,10,13-octahydro-1h,12h-benzo i pyrano 3,4-g indolizin-12-one hydrobromide, dh?e, dihydro-?-erythroidine hydrobromide, 3, a-1,6-didehydro-14,17-dihydro-3-methoxy-16 15h-oxaerythrinan-15-one hydrobromide, 3beta-1,6-didehydro-14,17-dihydro-3-methoxy-16 15h-oxaerythrinan-15-one hydrobromide
COC1CC=C2CCN3C2(C1)C4=C(CC3)COC(=O)C4.Br
11957537
>98%
50mg
C16H22BrNO3
GFIGWAJEIMHJJB-LINSIKMZSA-N
356.26
356.26
Videos