missing translation for 'onlineSavingsMsg'
Learn More

Dihexylamine, 99+%, Thermo Scientific Chemicals

Catalog Number p-3757713
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
143-16-8
C12H27N
185.35
MFCD00009521
PXSXRABJBXYMFT-UHFFFAOYSA-N
dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
8920
N-hexylhexan-1-amine
CCCCCCNCCCCCC
This item is not returnable. View return policy
143-16-8
C12H27N
185.35
MFCD00009521
PXSXRABJBXYMFT-UHFFFAOYSA-N
dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
8920
N-hexylhexan-1-amine
CCCCCCNCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 3°C
Color Colorless
Density 0.8000g/mL
Boiling Point 236°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (CH3(CH2)5)2NH
Refractive Index 1.4320 to 1.434
Quantity 5 g
UN Number 2927
Beilstein 04,I,384
Specific Gravity 0.8
Solubility Information Solubility in water: 0.3g/L(25°C)
Formula Weight 185.35
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dihexylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dihexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'