missing translation for 'onlineSavingsMsg'
Learn More

Diglycolic anhydride, 97%, Thermo Scientific Chemicals

Catalog Number p-111345
45.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4480-83-5
C4H4O4
116.07
MFCD00006677
PIYNUZCGMLCXKJ-UHFFFAOYSA-N
diglycolic anhydride, oxydiacetic anhydride, digylcolicanhydride, 4-maleimidobutyric acid hydrazide hydrochloride, r-1-methyl-2-pyrrolidineethanamine dihydrochloride, diglycolicanhydride, digylcolic anhydride, acetic acid, oxydi-, cyclic anhydride, p-dioxane-2,6-dione, pubchem24315
78232
1,4-dioxane-2,6-dione
C1C(=O)OC(=O)CO1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4480-83-5
C4H4O4
116.07
MFCD00006677
PIYNUZCGMLCXKJ-UHFFFAOYSA-N
diglycolic anhydride, oxydiacetic anhydride, digylcolicanhydride, 4-maleimidobutyric acid hydrazide hydrochloride, r-1-methyl-2-pyrrolidineethanamine dihydrochloride, diglycolicanhydride, digylcolic anhydride, acetic acid, oxydi-, cyclic anhydride, p-dioxane-2,6-dione, pubchem24315
78232
1,4-dioxane-2,6-dione
C1C(=O)OC(=O)CO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 90°C to 95°C
Color Cream to White
Boiling Point 240°C to 241°C
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Quantity 25 g
Beilstein 19, 153
Solubility Information Solubility in water: soluble
Formula Weight 116.07
Percent Purity 0.97
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material Diglycolic anhydride, 0.97
Show More Show Less
compliance-icons
Product Identifier
  • Diglycolic anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'