missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diglycolic anhydride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Diglycolic anhydride is used in the preparation of N,N-diethyldiglycollamsaeure by reacting with diethylamine. It is an useful bio chemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diglycolic anhydride is used in the preparation of N,N-diethyldiglycollamsaeure by reacting with diethylamine. It is an useful bio chemical for proteomics research.
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
Diglycolic anhydride is used in the preparation of N,N-diethyldiglycollamsaeure by reacting with diethylamine. It is an useful bio chemical for proteomics research.
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
Specifications
Melting Point | 91°C to 94°C |
Boiling Point | 240°C to 241°C |
Quantity | 10 g |
Beilstein | 112527 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in chloroform. |
Formula Weight | 116.07 |
Percent Purity | 0.97 |
Chemical Name or Material | Diglycolic anhydride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'