Learn More
Thermo Scientific™ Block, 1 x 96-well Elisa
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Brand: Thermo Scientific™ 88870111
Code : NEW
Additional Details : Weight : 1.70000kg
Description
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
- Warranty: 2 years
Specifications
1 x 96-well Elisa block | |
1 x 96-well Elisa | |
96 | |
76 x 124 x 39 mm (3 x 4.9 x 1.5 in.) |
Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models) | |
LED | |
Block |