Learn More
Grant Instruments™ Digital High Performance Refrigerated Baths
Brand:  Grant Instruments™ TX150-R2
Code : 57
Additional Details : Weight : 23.00000kg
Product Code. 12767829
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
-25 to 100°C
5L
110 x 145mm
19.2kg
Videos
Documentation