missing translation for 'onlineSavingsMsg'
Learn More

Difurfurylsulfide, 96%, ACROS Organics™

Brand:  Acros Organics 265080250

Code : 29

Additional Details : CAS Number : 13678-67-6 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11916171

  • £117.30 / 25g


Estimated Shipment
21-06-2018
Add to basket

Specifications

Chemical Identifiers

Difurfurylsulfide
96%
194.25
96%
C10H10SO2
2, 2'-Thiodimethylenedifurane

Specifications

Difurfurylsulfide
135.0° to 143.0°C (14.0 mmHg)
30.0° to 32.0°C
Authentic
Glass bottle
Low Melting Solid or Liquid
96%
194.25
>110°C
White to brown
25g
13678-67-6
1.5540 to 1.5560
96%
C10H10SO2
2, 2'-Thiodimethylenedifurane
Videos
SDS