missing translation for 'onlineSavingsMsg'
Learn More

Diethylphosphonoacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-109998
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3095-95-2
C6H13O5P
196.14
MFCD00192032
DVQMPWOLBFKUMM-UHFFFAOYSA-N
2-diethoxyphosphoryl acetic acid, diethylphosphonoacetic acid, diethyl carboxymethylphosphonate, diethoxyphosphorylacetic acid, diethyl phosphonoacetate, diethoxyphosphoryl acetic acid, acetic acid, diethoxyphosphinyl, acmc-20al82, diethylphosphonoethanoic acid, diethylcarboxymethylphosphonate
36704
2-diethoxyphosphorylacetic acid
CCOP(=O)(CC(=O)O)OCC
This item is not returnable. View return policy
3095-95-2
C6H13O5P
196.14
MFCD00192032
DVQMPWOLBFKUMM-UHFFFAOYSA-N
2-diethoxyphosphoryl acetic acid, diethylphosphonoacetic acid, diethyl carboxymethylphosphonate, diethoxyphosphorylacetic acid, diethyl phosphonoacetate, diethoxyphosphoryl acetic acid, acetic acid, diethoxyphosphinyl, acmc-20al82, diethylphosphonoethanoic acid, diethylcarboxymethylphosphonate
36704
2-diethoxyphosphorylacetic acid
CCOP(=O)(CC(=O)O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.2200g/mL
Boiling Point 150.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4440 to 1.4460
Quantity 5 g
Specific Gravity 1.22
Formula Weight 196.14
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material Diethylphosphonoacetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Diethylphosphonoacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'