Diethylmethoxyborane, 4M solution in THF, AcroSeal™, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 398381000

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 7397-46-8 Weight: 0.13130kg Transport: UN number : 3392 Chem class : 1 Pack group : I


Packaging AcroSeal™ Glass Bottle
Quantity 100mL

Product Code. 10225004

Quantity Price
1 £ 47.1 / 100mL
EU Stock 7
Add to basket

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Description and Specification

Chemical Identifiers

Chemical Name or Material Diethylmethoxyborane
Name Note 4M solution in THF
CAS 7397-46-8
CAS 109-99-9
Molecular Formula C5H13BO
Linear Formula CH3OB(CH2CH3)2
Formula Weight 99.97
MDL Number MFCD00013240

Specification

Formula Weight 99.97
Physical Form Solution
Quantity 100mL
Color Clear colorless to slightly amber
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3OB(CH2CH3)2
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact
CAS 7397-46-8
MDL Number MFCD00013240
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. May cause an allergic skin reaction. May cause respiratory irritation. May cause damage
Molecular Formula C5H13BO
Health Hazard 1 Danger
Density 0.8130g/mL
Name Note 4M solution in THF
Chemical Name or Material Diethylmethoxyborane
CAS 109-99-9
Documentation

Technical documents

Safety and Handling

Danger

GHS P Statement Causes severe skin burns and eye damage. Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. May cause an allergic skin reaction. May cause respiratory irritation. May cause damage

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact