Learn More
Thermo Scientific Acros Diethylenetriamine 98+% 0.9509g/mL m.p. -40°C b.p. 206°C (760) r.i. 1.4826 Acros Organics
Brand:  Thermo Scientific Acros 114310050
Diethylenetriamine 98+% 0.9509g/mL m.p. -40°C b.p. 206°C (760) r.i. 1.4826 Acros Organics
Code : 29
Additional Details : CAS Number : 111-40-0 Weight : 0.00480kg Transport : UN number : 2079 Chem class : 8 Pack group : II
Product Code. 10456720