missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol dibutyl ether, 99+%, Thermo Scientific Chemicals

Catalog Number p-7051870
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
112-73-2
C12H26O3
218.337
MFCD00009459
KZVBBTZJMSWGTK-UHFFFAOYSA-N
diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl
8210
1-[2-(2-butoxyethoxy)ethoxy]butane
CCCCOCCOCCOCCCC
This item is not returnable. View return policy
112-73-2
C12H26O3
218.337
MFCD00009459
KZVBBTZJMSWGTK-UHFFFAOYSA-N
diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl
8210
1-[2-(2-butoxyethoxy)ethoxy]butane
CCCCOCCOCCOCCCC

Extraction of precious metals

Extraction of precious metalsDiethylene glycol dibutyl ether is used as a solvent in Grignard reactions. It is also used as solvents in gold refining, decorative inks for ceramics and digital inks. It finds application in electrochemistry, gas absorption, extractant and high boiling reaction medium. It is also used in fuel, lubricant, textile and medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -60°C
Color Colorless
Density 0.885 g/mL
Boiling Point 256°C
Flash Point 117°C (243°F)
Refractive Index 1.4235
Quantity 25 mL
Solubility Information Miscible with dimethyl sulfoxide,ethanol and acetone. Immiscible with water.
Formula Weight 218.74
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Diethylene glycol dibutyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'