missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyanophosphonate, 93%, Thermo Scientific™

Catalog Number p-3778242
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2942-58-7
C5H10NO3P
163.11
MFCD00010256
ZWWWLCMDTZFSOO-UHFFFAOYSA-N
diethyl cyanophosphonate, diethyl phosphorocyanidate, diethylphosphorocyanidate, diethylcyanophosphate, diethyl cyanidophosphate, phosphorocyanidic acid, diethyl ester, diethoxy-phosphoryl cyanide, diethyl cyanophosphosphonate, cyanophosphonic acid diethyl ester, diethylcyanophosphonate
76257
CCOP(=O)(OCC)C#N
This item is not returnable. View return policy
2942-58-7
C5H10NO3P
163.11
MFCD00010256
ZWWWLCMDTZFSOO-UHFFFAOYSA-N
diethyl cyanophosphonate, diethyl phosphorocyanidate, diethylphosphorocyanidate, diethylcyanophosphate, diethyl cyanidophosphate, phosphorocyanidic acid, diethyl ester, diethoxy-phosphoryl cyanide, diethyl cyanophosphosphonate, cyanophosphonic acid diethyl ester, diethylcyanophosphonate
76257
CCOP(=O)(OCC)C#N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 104.0°C to 105.0°C (19.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CN
Refractive Index 1.4020 to 1.4040
Quantity 5 g
Fieser 05,17; 06,192; 07,107; 10,145; 15,26; 16,24
Specific Gravity 1.07
Solubility Information Solubility in water: decomposes in water
IUPAC Name diethoxyphosphorylformonitrile
Formula Weight 163.11
Percent Purity 93%
Physical Form Liquid
Chemical Name or Material Diethyl cyanophosphonate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'