missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl cyanophosphonate, 93%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for C-C Bond Formation (Olefination)
Specifications
Color | Colorless to Yellow |
Density | 1.0700g/mL |
Boiling Point | 104.0°C to 105.0°C (19.0 mmHg) |
Flash Point | 80°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 91% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (C2H5O)2P(O)CN |
Refractive Index | 1.4020 to 1.4040 |
Quantity | 5 g |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'