missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyanophosphonate, 93%, Acros Organics™

Brand:  Acros Organics 224290250

Code : 29

Additional Details : CAS Number : 2942-58-7 Weight : 0.07500kg Transport : UN number : 2922 Chem class : 1a Pack group : II

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10581471

  • £120.50 / 25g


Estimated Shipment
16-03-2018
Add to basket

Specifications

Chemical Identifiers

Diethyl cyanophosphonate
93%
(C2H5O)2P(O)CN
MFCD00010256
93%
C5H10NO3P
163.11
Diethylphosphoryl cyanide

Specifications

Diethyl cyanophosphonate
Authentic
1.4020 to 1.4040
Clear colorless to yellow
Liquid
93%
80°C
(C2H5O)2P(O)CN
163.11
Diethylphosphoryl cyanide
2942-58-7
Glass bottle
1.0700g/mL
104.0° to 105.0°C (19.0 mmHg)
93%
25g
C5H10NO3P
1.07
MFCD00010256
Videos
SDS