missing translation for 'onlineSavingsMsg'
Learn More

Diethyl chloromalonate, 90%, Thermo Scientific Chemicals

Catalog Number p-4258949
Click to view available options
Quantity:
5 mL
25 mL
14064-10-9
C7H11ClO4
194.61
WLWCQKMQYZFTDR-UHFFFAOYSA-N
diethyl chloromalonate, diethyl 2-chloromalonate, ethyl chloromalonate, ethylmonochloromalonate, propanedioic acid, chloro-, diethyl ester, chloromalonic acid, diethyl ester, diethyl chloropropanedioate, malonic acid, chloro-, diethyl ester, 1,3-diethyl 2-chloropropanedioate, propanedioic acid, 2-chloro-, 1,3-diethyl ester
84182
diethyl 2-chloropropanedioate
CCOC(=O)C(C(=O)OCC)Cl
14064-10-9
C7H11ClO4
194.61
WLWCQKMQYZFTDR-UHFFFAOYSA-N
diethyl chloromalonate, diethyl 2-chloromalonate, ethyl chloromalonate, ethylmonochloromalonate, propanedioic acid, chloro-, diethyl ester, chloromalonic acid, diethyl ester, diethyl chloropropanedioate, malonic acid, chloro-, diethyl ester, 1,3-diethyl 2-chloropropanedioate, propanedioic acid, 2-chloro-, 1,3-diethyl ester
84182
diethyl 2-chloropropanedioate
CCOC(=O)C(C(=O)OCC)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 221.0°C to 222.0°C
Color Colorless
Density 1.2040g/mL
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Linear Formula C2H5O2CCH(Cl)CO2C2H5
Refractive Index 1.431
Quantity 5 mL
Specific Gravity 1.204
Formula Weight 194.61
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Diethyl chloromalonate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethyl chloromalonate
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'