missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-oxopropyl)phosphonate, 96%, Thermo Scientific Chemicals

Catalog Number 10607294
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1067-71-6
C7H19O4P
198.2
MFCD00044728
RSAFKRSMGOSHRK-UHFFFAOYSA-N
diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate
14028
1-diethoxyphosphorylpropan-2-one
CCOP(=O)(CC(=O)C)OCC
This item is not returnable. View return policy
1067-71-6
C7H19O4P
198.2
MFCD00044728
RSAFKRSMGOSHRK-UHFFFAOYSA-N
diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate
14028
1-diethoxyphosphorylpropan-2-one
CCOP(=O)(CC(=O)C)OCC

For C-C Bond Formation (Olefination)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 98.0°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CH3COCH2P(=O)(OCH2CH3)2
Refractive Index 1.4325 to 1.4355
Quantity 25 g
Fieser 13,255
Specific Gravity 1.11
Formula Weight 198.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Diethyl (2-oxopropyl)phosphonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'