missing translation for 'onlineSavingsMsg'
Learn More

Diethylcarbamyl Chloride 99%, Thermo Scientific Chemicals

Catalog Number 10520401
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
88-10-8
C5H10ClNO
135.591
MFCD00000636
OFCCYDUUBNUJIB-UHFFFAOYSA-N
diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride
6916
N,N-diethylcarbamoyl chloride
CCN(CC)C(=O)Cl
This item is not returnable. View return policy
88-10-8
C5H10ClNO
135.591
MFCD00000636
OFCCYDUUBNUJIB-UHFFFAOYSA-N
diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride
6916
N,N-diethylcarbamoyl chloride
CCN(CC)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -32.0°C
Boiling Point 117.0°C to 123.0°C (132.0 mmHg)
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula (C2H5)2NCOCl
Beilstein 04, 120
Packaging Glass Bottle
Refractive Index 1.4505 to 1.4525
Formula Weight 135.59
Percent Purity 99%
Density 1.0700g/mL
Quantity 100 mL
Flash Point 80°C
Specific Gravity 1.07
Physical Form Liquid
Chemical Name or Material Diethylcarbamyl chloride, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'