missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylcarbamyl Chloride 99%, Thermo Scientific Chemicals
16.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -32.0°C |
Boiling Point | 117.0°C to 123.0°C (132.0 mmHg) |
Assay Percent Range | 99% |
Infrared Spectrum | Authentic |
Linear Formula | (C2H5)2NCOCl |
Beilstein | 04, 120 |
Packaging | Glass Bottle |
Refractive Index | 1.4505 to 1.4525 |
Formula Weight | 135.59 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'