missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diethylamine hydrobromide, 98%

Brand:  Alfa Aesar™ A10452.0B

Code : NEW

Additional Details : CAS Number : 6274-12-0 Weight : 1.00000kg

Quantity 1000g

 View more versions of this product

Product Code. 11498663

  • £139.50 / 1000g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Diethylamine hydrobromide
(CH3CH2)2NH·HBr
MFCD00012498
Diethylammonium bromide
6274-12-0
154.06
3905681

Specifications

Diethylamine hydrobromide
6274-12-0
(CH3CH2)2NH·HBr
MFCD00012498
218° to 220°C
1000g
98%
154.06
3905681
Diethylammonium bromide
Videos
SDS