Learn More
Alfa Aesar™ Diethylaluminum ethoxide, 25% w/w in hexane
Brand: Alfa Aesar™ 087625.NC
Code : NEW
Additional Details : CAS Number : 1586-92-1 Weight : 0.01000kg Transport : UN number : 3394 Chem class : 1 Pack group : I
Quantity | 10g |
---|
Chemical Identifiers
Diethylaluminum ethoxide | |
C6H15AlO | |
130.17 | |
UN3394 |
1586-92-1 | |
(C2H5)2Al(OC2H5) | |
MFCD00009059 |
Specifications
Diethylaluminum ethoxide | |
1586-92-1 | |
−23°C (−9°F) | |
Colorless | |
Air & Moisture Sensitive | |
C6H15AlO | |
130.17 | |
UN3394 |
25% w/w in hexane | |
10g | |
0.851g/mL | |
Petrol-like | |
Liquid | |
(C2H5)2Al(OC2H5) | |
MFCD00009059 |