Learn More
Thermo Scientific Alfa Aesar Diethylaluminum chloride, 96%, 25% w/w in heptane
Brand: Thermo Scientific Alfa Aesar 089035.NB
Diethylaluminum chloride, 96%, 25% w/w in heptane
Code : NEW
Additional Details : CAS Number : 96-10-6 Weight : 0.00500kg Transport : UN number : 3394 Chem class : 4.2 Pack group : I