missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl pyrocarbonate, 97%, pure, ACROS Organics™
Click to view available options
2.5kg
Specifications
CAS | 1609-47-8 |
Density | 1.1175g/mL |
Boiling Point | 93.0°C to 94.0°C (18.0mmHg) |
Flash Point | 69°C |
Assay Percent Range | 0.97 |
Molecular Formula | C6H10O5 |
Linear Formula | O(CO2C2H5)2 |
MDL Number | MFCD00009106 |
Beilstein | 03,IV,18 |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- Diethyl pyrocarbonate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'